Shoe covers, PP, Hopen® Safefeet® SkidGuard™, with PE sole
Lieferant: KOLMI HOPEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10.007
113-9989EA 170 EUR InStock
113-9989
Shoe covers, PP, Hopen® Safefeet® SkidGuard™, with PE sole
Einweg-Kleidung
PP shoe covers feature a 60 μm PE plastic-coated, non-slip sole to prevent slipping and ensure a long product life.
  • Class 1 medical device
  • Single use, non-sterile
  • PP non-woven upper with PE sole
  • Made in France
These lightweight garments are adaptable in size to fit over all types of shoes. The PE sole helps to reduce the risk of abrasion on rough floors.
Ideal for use in medical and clinical areas, for food processing, and in laboratories.
Sole thickness: 60 μm
Weight: 35 g/m²
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge