Safety shoes, lace-up, Triaxe® Urban TXL Flyer Ripper
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6244457/45 6244457/46 6244457/47 6244457/39 6244457/38 6244457/40 6244457/41 6244457/42 6244457/43 6244457/44
114-0649EA 0 EUR InStock
114-0649 114-0650 114-0651 114-0643 114-0642 114-0644 114-0645 114-0646 114-0647 114-0648
Safety shoes, lace-up, Triaxe® Urban TXL Flyer Ripper
Schuhe und Stiefel
Durable safety shoe with a water resistant full grain leather upper and Cordura® Flexible reinforcements over the front. The shoe was specially adapted to employees in cleaning companies and local authorities.
  • 200 J FREELIUM™ Toe cap: Insulating, light and non-metallic
  • Ergonomical, anti-bacterial insole: Removable and changeable for an optimal comfort
  • TRIAXE™ Urban PU outsole: shock absorbent
EN ISO 20345:2004/AM 1:2007; S3, CI, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge