Sicherheitsschuhe, zum Schnüren, The Roar, Trophy
Lieferant: U-Power
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SN20056/35 SN20056/37 SN20056/36 SN20056/38 SN20056/39 SN20056/40 SN20056/41 SN20056/42 SN20056/43 SN20056/44 SN20056/45 SN20056/46 SN20056/47 SN20056/48
114-2798EA 96.4 EUR InStock
114-2798 114-2800 114-2799 114-2813 114-2814 114-2815 114-2816 114-2817 114-2818 114-2819 114-2820 114-2833 114-2834 114-2835
Sicherheitsschuhe, zum Schnüren, The Roar, Trophy
Schuhe und Stiefel
Safety shoes made from extra strong breathable canvas with soft suede leather inserts and a slip-resistant dual-density PU sole.
  • Light non-slip Action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Dark blue
Colour: Dark blue
Zertifizierungen: EN 20345:2011; S3, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge