Sicherheitsschuhe, zum Schnüren, The Roar, King
Lieferant: U-Power
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SN20014/38 SN20014/39 SN20014/40 SN20014/41 SN20014/42 SN20014/43 SN20014/44 SN20014/45 SN20014/46 SN20014/47 SN20014/48 SN20014/35 SN20014/36 SN20014/37
114-2867EA 104.8 EUR InStock
114-2867 114-2868 114-2869 114-2870 114-2871 114-2872 114-2873 114-2874 114-2875 114-2876 114-2877 114-2864 114-2865 114-2866
Sicherheitsschuhe, zum Schnüren, The Roar, King
Schuhe und Stiefel
Safety shoes made from soft water repellent leather and a slip-resistant dual-density PU sole.
  • Light non-slip Action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Zertifizierungen: EN 20345:2011; S3, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge