Sicherheitsschuhe, Slipper, Bacou Pro'Clean
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6246127/39
114-3339EA 89.4 EUR InStock
114-3339
Sicherheitsschuhe, Slipper, Bacou Pro'Clean
Schuhe und Stiefel
These safety shoes are made from breathable water resistant microfibre upper and a dual-density PU sole (resistant to abrasion, fuel oil and grease).
  • Microfibre upper for maximum comfort and lightness
  • Removable, antibacterial and anti-static, dual-density PU foam insole with comfortable heel pads
  • No stitching on the upper, limits contamination; easy care
  • Light and flexible, slip resistant and self-cleaning, dual-density PU outsole ensures shock absorption and protects against changes in temperature
  • Light, metal-free, Spacium™ toecap in injected polymer (200 J)
Ideal for use in catering, food industries, industrial cleaning laboratories plus medical and pharmaceutical industries.
Zertifizierungen: EN ISO 20345; S2, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge