ESD-Sicherheitsschuhe, Velcro®,TX 360, Atlas
Lieferant: ATLAS - the shoe company
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
55145 55136 55137 55138 55139 55140 55141 55142 55143 55146 55147 55148 55149
114-4679EA 0 EUR InStock
114-4679 114-4670 114-4671 114-4672 114-4673 114-4674 114-4675 114-4676 114-4677 114-4680 114-4681 114-4682 114-4683
ESD-Sicherheitsschuhe, Velcro®,TX 360, Atlas
Schuhe und Stiefel
Lightweight ESD safety shoe made from perforated Sportline upper (oiled nubuck leather), breathable functional lining and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: Pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Farbe: Schwarz
Zertifizierungen: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge