Protect (i)Xtreme Safety Ankle Boots, Lace-Up
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6246204/38 6246204/39 6246204/40 6246204/41 6246204/42 6246204/43 6246204/44 6246204/45 6246204/46 6246204/47 6246204/48
114-5903EA 187 EUR InStock
114-5903 114-5894 114-5895 114-5893 114-5892 114-5891 114-5890 114-5889 114-5888 114-5883 114-5881
Protect (i)Xtreme Safety Ankle Boots, Lace-Up
Schuhe und Stiefel
Non metallic full grain, leather ankle boot with a strong water resistant upper construction and an innovative PU/nitrile outsole for extrem performances.
  • (i)NTELLIGENT WEAR CONTROL: Helps users to detect when the outsole is used
  • Bi-density insole for improved comfort
  • Non metallic anti-puncture midsole
  • Smartium non metallic toecap made from injected polymer
Colour: Black.
Zertifizierungen: EN ISO 20345:2011; S3, SRC, HI, HRO, CI.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge