VWR®, Cleanroom clogs
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
114-8980EA 170 EUR InStock
114-8980 114-8981 114-8982 114-8983 114-8984 114-8985
VWR®, Cleanroom clogs
Schuhe und Stiefel
VWR® cleanroom clogs provide comfort and durability.
  • Anti-static
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non slip sole
  • Anatomic heel basin
They have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day extended comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilisation. They can also be cleaned in 100 °C with domestic detergents, liquid disinfecting solutions, EO, or formaldehyde.
Ideal for use with VWR® cleanroom disposable or launderable socks.
Bestellinformation: Clogs can be ordered by the pair or per case.
Verpackung: Packed six pairs per case.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge