ESD safety shoes, BRAYDEN XXTM, strap closure, low, S3
Lieferant: ELTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
720531/36 720531/37 720531/38 720531/39 720531/40 720531/41 720531/42 720531/43 720531/44 720531/45 720531/46 720531/47 720531/48
114-9127EA 88.9 EUR InStock
114-9127 114-9128 114-9129 114-9130 114-9131 114-9132 114-9133 114-9134 114-9135 114-9136 114-9137 114-9138 114-9139
ESD safety shoes, BRAYDEN XXTM, strap closure, low, S3
Schuhe und Stiefel
Excellent wearer comfort, with a padded upper edge to protect the Achilles’ tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO grey
  • Non metallic, penetration resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
Excellent discharge capability, the shoe is suitable for work in ESD-sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Metal- and leather-free.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge