Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bromkresolgrün Natriumsalz in ACS analysereinem Wasser ≥90% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ200

114367-25G 114367-5G
114367-25GEA 285 EUR InStock
114367-25G 114367-5G
Bromkresolgrün Natriumsalz in ACS analysereinem Wasser ≥90% (durch HPLC), Sigma-Aldrich®
Bromkresolgrün Natriumsalz
Bromocresol Green (BCG), also known as bromcresol green, is a synthetic, anionic, triphenylmethane dye belonging to the phthalein and sulphonphthalein dyes group. It is commonly used as a pH indicator as it changes colour from yellow to blue-green at pH 3,8 to 5,4. The monosodium salt of bromocresol green is highly ionisable in water. The dye at a higher pH level produces a large and weakly lipophilic dianion.

  • Bromocresol green serves as a pH indicator in biology and medicinal experiments
  • It is used widely as a vital stain to study blood-brain permeability
  • It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations, and in charge-transfer complexation processes
  • It is used in TLC for the visualisation of compounds with functional groups whose pKa is below 5
  • It has been used in the pH-sensitive colorimetric determination of glutamate decarboxylase activity
  • It has also been employed in the evaluation of paper-analytical devices for colorimetric analysis of serum samples
  • It is suitable as a selective inhibitor in kidney physiology studies
  • It is a diagnostic assay manufacturing, hematology, histology


Formel: C₂₁H₁₃Br₄NaO₅S
Melting Pt: 230 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00148898
CAS-Nummer: 62625-32-5
EINECS: 263-657-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6576f0c57895b913ac2b0cdaad5ae705df1a91ad&vid=6576f0c57895b913ac2b0cdaad5ae705df1a91ad&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Visual Interval 1, pH 3.8 = Yellow Pass
Visual Interval 2, pH 4.5 = Green Pass
Visual Interval 3, pH 5.4 = Blue Pass
Clarity of solution not more than a faint trace of turbidity or insoluble matter c = 0.1 g/100 ml; Water Conforms
Purity (HPLC) ≥90 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge