Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Methylorange in ACS analysereinem Wasser 85% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

114510-25G 114510-100G 114510-500G
114510-25GEA 36 EUR InStock
114510-25G 114510-100G 114510-500G
Methylorange in ACS analysereinem Wasser 85% (Farbstoffgehalt), Sigma-Aldrich®
Methylorange
Methyl orange (C.I. 13025) is the sodium salt of p-dimethylaminoazobenzenesulfonic acid. It is an orange-colored synthetic dye, which changes color depending on the pH of the solution. It develops a red color in acidic solution and yellow color in basic solution.

  • Methyl orange is used as a:
  • pH indicator in the cell sap
  • Counterstain to crystal violet in staining pollen tubes
  • Component of a polychrome histological stain
  • Solution indicator, such as in assays of oxalic and malonic acids
  • In other biomedical applications that include use in film dosimeters and as a reagent for the assay of bromide ions


Formel: C₁₄H₁₄N₃NaO₃S
Siedepunkt: ∼90 °C (1013 hPa)
Melting Pt: >300 °C
Dichte: ∼1 g/cm³ (20 °C)
Flash-Pt: 38 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00007502
CAS-Nummer: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 13,06127
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=915f043445c2be992ab36bc290d60762365a46ad&vid=915f043445c2be992ab36bc290d60762365a46ad&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (colour) Orange
Appearance (form) Powder
Infrared spectrum Conforms to structure
Visual interval, pH 3.0 pink or red Pass
Visual interval, pH 3.2 pinkish orange Pass
Visual interval, pH 4.4 yellow Pass
Appearance (clarity) Pass
Carbon 43.7 - 59.1%
Nitrogen 10.9 - 14.8%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge