11488.36
11488.A3
11488.36EA
58
EUR
InStock
11488.36
11488.A3
Ammoniumtetrafluoroborat ≥97%
Ammonium tetrafluoroborate
Purity: 97%
CAS No.: 13826-83-0
Molecular Formula: NH4BF4
Formula Weight: 104.84
Physical Form: Powder
Density: 1.871
Appearance: White
Odour: Ammonia-like
Melting Point: 220° subl.
UN No.: 3260
MDL No.: MFCD06245785