Reinraum-Wischtücher, Amplitude™ Prozorb®
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PZ-99 PZ-1212
115-0272EA 487 EUR InStock
115-0272 CONTPZ-1212
Reinraum-Wischtücher, Amplitude™ Prozorb®
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Amplitude™ Prozorb® wipes are a hydroentangled blend of cellulose and polyester, produced through a special process that creates apertures in the fabric while enhancing the thickness and providing excellent cleanliness.
  • Durable textured surface promotes cleaning and particle removal capability
  • Excellent sorbency, great level of intrinsic sorbency and quick rate of sorbency
  • Low level of particles and extractables
Ideal for spill clean up in critical environments, biofilm and residue removal, product wipe down prior to cleanroom entry and wiping to dry.
Recommended for ISO Class 5 - 8 environments.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge