Aerospace cleaning wipes, Sontara® AC™
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AC12165
115-0370EA 155 EUR InStock
115-0370
Aerospace cleaning wipes, Sontara® AC™
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für sensible Anwendungen
Sontara AC is a white non woven creped wipe made with 55% special cellulose and 45% polyester.
  • Extremely low lint: Leave surfaces clean
  • High strength, good absorbency and versatility
  • Contain no binders, solvent resistant
  • Durable: Wipes maintain their strength, even when wet
Ideal for a wide range of critical cleaning tasks essential to industries such as biotech, pharmaceutical and electronics, they are very effective in applications such as surface preparation for coatings, machining operations, and general maintenance. Also suitable for pre-painting tasks.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge