Wipe rolls, Multitex® DR
Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11830-02
115-2743EA 150 EUR InStock
115-2743
Wipe rolls, Multitex® DR
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
These reusable wipes made from viscose fleece are suitable for disinfecting and cleaning applications. They are intended to be used with the Wiper Bowl® Safe and Clean dispenser bucket.
  • Perforated roll
  • Tear-resistant
  • Highly absorbent and lint-free
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge