Reinraum-Wischtücher, Polynit-Z
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LWZS0001
115-2953EA 913 EUR InStock
115-2953
Reinraum-Wischtücher, Polynit-Z
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
The Polynit-Z wipe features conductive yarns within the polyester interlock knit. It is a very clean static dissipative wipe that is well suited for applications that are sensitive to electrostatic discharge. The average surface resistivity is 3,5×10¹⁰ ohms/sq.
  • Very clean sealed edges that are non abrasive
  • Laundered and packaged in an ISO Class 4 cleanroom
  • Low in both particles and extractables
Zur Verwendung in Reinräumen der ISO-Klassen 3 - 4 geeignet.
Farbe: Weiß
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge