Heel grounder, Wescorp
Lieferant: DESCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
249265
115-3062EA 7.7 EUR InStock
115-3062
Heel grounder, Wescorp
Erdung ESD-Fersen- und Zehenbänder
Wescorp heel ground with one Megohm resistor are designed to satisfy Footwear requirements of IEC 61340-5-1.
  • Wear resistant rubber
  • Provides contact area to floor
  • Non-marking interior
  • Will not mark shoes
  • Snap-loc fastening assembly
  • Genuine non-fraying Hook and Loop material - adjustable and comfortable
  • 30,5 cm long, 8 strand nylon polyester grounding tab
  • Does not particulate; superior body to ground contact
  • Rg: 1×10⁶ < ×10⁷ Ω per IEC 61340-5-1 Clause A.2
Per IEC 61340-5-2 paragraph 5.2.8, footwear should be used 'where personnel are mobile, in storing areas, operating large machinery, etc., it may be impractical or dangerous to have long cords attached to them.'
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge