Conical earth bonding point cord
Lieferant: DESCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60474
115-3069EA 20.5 EUR InStock
115-3069
Conical earth bonding point cord
Erdung Erdungsleitungen
This cord with low profile helps keep employees from tripping on cord connection.
  • 10 mm snap stud easily snaps to mat socket, flush to mat surface
  • Extra ring terminal provided for customising cord length
  • Ring terminal at end of cord allows easy attachment to building ground
Stud has threaded hole; 6-32×6,35 mm flat head screw and countersunk washer are provided which allows ground cord to be bolted to mat; keeps cord from disconnecting; ensures proper grounding.
Lieferumfang: Includes 1 Megohm resistor.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge