Cleaning cloths, Tork, W8
Lieferant: Essity
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
510150
115-3121EA 158 EUR InStock
115-3121
Cleaning cloths, Tork, W8
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
This soft and flexible multipurpose cloth made of chemical pulp, polyester and PP quickly absorbs liquids and oil. The cloth provides a professional cleaning result thanks to the exelCLEAN™ effect.
  • Long-lasting cloth suitable for repeated use in wet & dry conditions
  • Efficient absorb and release properties mean less product needed to do the same job
  • Single wipe dispensing in portable and small size format for compact spaces
Food contact approved.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge