Silicone bench protector
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
350002
115-9137EA 162 EUR InStock
115-9137
Silicone bench protector
Oberflächenschutz Oberflächenschutz
Silicone sheet designed for laboratory benchtop protection or for use in autoclaves to protect any material.
  • Easy to clean with a wet cloth
  • Heat resistant up to 260 ºC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge