Magbeads FastRNA kit, 96 preparations, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
116572096
116572096.EA 767 EUR InStock
116572096.
Magbeads FastRNA kit, 96 preparations, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
MagBeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples.
  • High quality RNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals
MagBeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples and is based on superparamagnetic particles purification technology, no phenol-chloroform extraction or alcohol precipitation. The whole extraction process is only 60 minutes.
Purified RNA is ready for downstream applications such as RTPCR, virus RNA testing and so on. The kits buffers can be used for both manual extraction process and automatic nucleic acid extraction machines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge