11834.14
11834.22
11834.88
AB202578.0025
11834.14EA
70.7
EUR
InStock
11834.14
11834.22
11834.88
ABCRAB202578.0025
Hafnium(IV)chlorid ≥98% (Metall-Basis ausgeschlossen Zr) (max. 2,7% Zr)
Hafnium(IV)chlorid
Purity: 98+%
CAS No.: 13499-05-3
Molecular Formula: Cl4Hf
Formula Weight: 320.30
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Odour: Irritating
Melting Point: 320° subl.
Merck Reference: 14,4588
UN No.: 3260
MDL No.: MFCD00011034