11837.22
11837.36
11837.A3
11837.22EA
71.6
EUR
InStock
11837.22
11837.36
11837.A3
Molybdän(VI)oxid ≥99,95% (Metall-Basis)
Molybdenum(VI) oxide
Purity: 99.95%
CAS No.: 1313-27-5
Molecular Formula: MoO3
Formula Weight: 143.94
Physical Form: Sublimed Powder
Density: 4.692
Appearance: White to grey
Odour: Odourless
Melting Point: 795°
Boiling Point: 1155° subl.
Merck Reference: 14,6239
UN No.: 3288
MDL No.: MFCD00003469