11868.36
11868.A3
11868.A9
11868.36EA
41
EUR
InStock
11868.36
11868.A3
11868.A9
Mangan(II)chlorid, wasserfrei 97%
Mangan(II)chlorid
Purity: 97%
CAS No.: 7773-01-5
Molecular Formula: Cl2Mn
Formula Weight: 125.84
Physical Form: Crystalline/Flake
Density: 2.977
Appearance: Pink
Odour: Odourless
Melting Point: 650°
Boiling Point: 1190°
Merck Reference: 14,5728
UN No.: 3288
MDL No.: MFCD00011114