11890.03
11890.06
11890.03EA
278
EUR
InStock
11890.03
11890.06
Dinatriumhexachloroiridat Hexahydrat (≥33,9% Ir)
Dinatriumhexachloroiridat Hexahydrat
CAS No.: 19567-78-3
Molecular Formula: Na2IrCl6.6H2O
Formula Weight: 558.99 (450.92anhy)
Physical Form: Powder
Appearance: Red-black
Melting Point: 600° dec.
MDL No.: MFCD00149169