Drum wrench
Lieferant: JUSTRITE MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
08805
119-0238EA 55.8 EUR InStock
119-0238
Drum wrench
Schraubenschlüssel
Drum Wrench opens drum bung caps.
  • Rugged and non-sparking
  • Design of handle protects the hand
This corrosive-resistant tool fits many 50,8 or 19,05 mm bung styles commonly used on drums.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge