Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Economy spill kit

Lieferant: Brady

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

813858 813857
121-0190EA 69.6 EUR InStock
121-0190 121-0189
Economy spill kit
Absorptionsmaterial
Economy spill kit for chemicals are designed for small spills of chemicals and hazardous liquids while kit for maintanance are designed for small spills of industrial liquids including oils, water, solvents, coolants. This kit is ideal for transport of small quantities of liquids.

  • Compact in size
  • Requires small storage area
  • Water-resistant
  • Lightweight bag keeps sorbents dry

Lieferumfang: Supplied with 10 pads (410×510 mm), 2 socs (Ø 76×1220 mm), one pair of gloves and one disposable bag.

Verpackung: Packed in a yellow PVC bag with handles.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge