12232.14
12232.30
12232.A1
12232.14EA
39.4
EUR
InStock
12232.14
12232.30
12232.A1
Strontiumnitrat ≥99,0% ACS
Strontium nitrate
Purity: 99.0% min
CAS No.: 10042-76-9
Molecular Formula: N2O6Sr
Formula Weight: 211.63
Physical Form: Crystalline
Density: 2.990
Appearance: White
Odour: Odourless
Melting Point: 570°
Merck Reference: 14,8845
UN No.: 1507
MDL No.: MFCD00011248