12313.A3
12313.36
12313.A9
12313.A3EA
157
EUR
InStock
12313.A3
12313.36
12313.A9
Zink(II)nitrat Hexahydrat ≥99% (Metall-Basis)
Zink(II)nitrat Hexahydrat
Purity: 99%
CAS No.: 10196-18-6
Molecular Formula: N2O6Zn.6H2O
Formula Weight: 297.47 (189.38anhy)
Physical Form: Crystalline
Density: 2.07
Appearance: White
Odour: Odourless
Melting Point: ca 36°
Solubility: Soluble in water, alcohol
Merck Reference: 14,10144
UN No.: 1514
MDL No.: MFCD00149889