12360.14
12360.30
12360.A1
12360.A7
12360.14EA
38.4
EUR
InStock
12360.14
12360.30
12360.A1
12360.A7
Aluminiumnitrat Nonahydrat ≥98%
Aluminiumnitrat Nonahydrat
Purity: 98%
CAS No.: 7784-27-2
Molecular Formula: Al(NO3)3.9H2O
Formula Weight: 375.13 (212.99anhy)
Physical Form: Crystals
Sensitivity: Hygroscopic
Merck Reference: 14,352
UN No.: 1438
MDL No.: MFCD00003419