12362.36
12362.A1
12362.A7
12362.36EA
42
EUR
InStock
12362.36
12362.A1
12362.A7
Aluminiumsulfat Hydrat ≥97%
Aluminiumsulfat Hydrat
Purity: 97+%
CAS No.: 17927-65-0
Molecular Formula: Al2(SO4)3.xH2O (x~14-18)
Formula Weight: 342.15(anhy)
Physical Form: Crystalline
Density: 1.61
pH: 3.5
Merck Reference: 14,366
MDL No.: MFCD00149136