12411.36
12411.A3
12411.B4
12411.A9
12411.36EA
39.6
EUR
InStock
12411.36
12411.A3
12411.B4
12411.A9
1,2,4-Trimethylbenzol ≥98%
1,2,4-Trimethylbenzol
Purity: 98%
CAS No.: 95-63-6
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 48°(118°F)
Physical Form: Liquid
Density: 0.882
Refractive Index: 1.5040
Odour: Aromatic
Melting Point: -44°
Boiling Point: 169-171°
Solubility: Soluble in alcohol, benzene, ether
Beilstein Reference: 1903005
Merck Reference: 14,7915
UN No.: 3295
MDL No.: MFCD00008527