12610.22
12610.36
12610.22EA
32.4
EUR
InStock
12610.22
12610.36
Kaliumchromat ≥99,0% ACS
Potassium chromate
Purity: 99.0% min
CAS No.: 7789-00-6
Molecular Formula: K2CrO4
Formula Weight: 194.19
Physical Form: Crystalline
Density: 2.73
Appearance: Yellow
Odour: Odourless
Melting Point: 975°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7622
UN No.: 3087
MDL No.: MFCD00011368
In finishing leather, rust proofing of metals, textile mordant, as an analytical lab reagent