12832.03
12832.06
12832.03EA
81.2
EUR
InStock
12832.03
12832.06
Quecksilberselenid ≥99,999% (Metall-Basis)
Quecksilberselenid
Purity: 99.999%
CAS No.: 20601-83-6
Molecular Formula: HgSe
Formula Weight: 279.55
Physical Form: -100 Mesh Powder
Density: 8.21
Appearance: Gray
Odour: Odourless
Melting Point: subl.
UN No.: 2025
MDL No.: MFCD00049743