Mopps, AlphaMop™, PE
Lieferant: Texwipe
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TX7108 TX7118 TX7026 STX7118 TX7118M TX7118F TX7108H TX7123 STX7118M
129-0180EA 200 EUR InStock
129-0180 129-0181 141-0249 141-1048 141-1060 141-1059 141-1057 141-1064 141-1049
Mopps, AlphaMop™, PE
Mopps
Ideal for cleaning ceilings, walls, floors, and other flat surfaces in critical environments. The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fibreglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. A lightweight, durable fibreglass handle gives the user excellent control over the swivel head.
  • Lightweight mop maintains ultra-low generation of particles and extractables
  • Mop can be sterilised
  • Mophead covers can be pre-wetted with 6% isopropanol
  • Accesses hard-to-reach areas with ease
  • Excellent durability with low contamination
  • Allows user to change covers and foam pads frequently, adhering to wiping protocol
  • Rapid-change cover and foam pad configuration
* Handle length.
Verpackung: Mopp ist reinraumgerecht Bag-Within-A-Bag® verpackt.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge