Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Schwämme, NovaPoly™

Lieferant: MICRONOVA

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

SP4-533 SP68-46 SP68-745 SP20-46 SP6-363 SP68-46IR SPAA-363 SP6-745 SP2-46 SP6-33 SP68-33 SP6-46
129-0639EA 667 EUR InStock
129-0639 129-0641 115-1877 129-0638 MICRSP6-363 MICRSP68-46IR 115-1815 MICRSP6-745 MICRSP2-46 115-1817 MICRSP68-33 MICRSP6-46
Schwämme, NovaPoly™
Wischtücher Schwämme
Cleanroom sponges to meet every demand from isolators and laminar flow benches to equipment interiors and laboratory applications. Choose from a range of fabric covers for specialized cleaning.

  • Made with cleanroom approved materials
  • Durable, reusable and absorbent
  • Fabric covers securely sewn over reticulated urethane foam interiors
  • Shape and size options available
  • Available gamma irradiated for aseptic areas
  • Hold up to strong disinfectants
  • Autoclavable

NovaCel™ Sponge: Ultra-clean polyvinyl acetate, firm to the touch, but soft and pliant once wetted - ideal for lapping and polishing in wafer manufacture.

MegaTex® Sponge: Textured, thermally bonded, non-woven polyamide for textured surfaces, heavy soils, and viscous spills.
PolyMesh™ Sponges: 100% polyester with monofilament mesh - provides increased abrasion to remove photo resist, chemical deposits, and fluxes. Also used for fermentation tanks and veterinary laboratories.

NovaPoly™ Sponges: 100% smooth knit polyester - excellent disinfectant dispersal for a wide variety of cleanroom applications; FDA approved for indirect food contact.
NovaLite™ Sponges: latex bonded with rayon, chamois material, with 400% absorbency and great friction resistance - ideal for removing dust and fine powders in pharmaceutical, medical device and hi-tech cleanrooms. Excellent alternative to PVA where autoclaving is one of the criteria.

Microfiber Sponges: 80% polyester/ 20% polyamide material with superior cleaning ability and over 400% absorbency.
NovaSponges™: bonded, non-woven polyvinyl alcohol (PVA) - Soft and pliable, yet durable and hard wearing for wet or dry applications. Caution: Not recommended for use with isopropyl alcohol or chlorides.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge