Sampling bag, sterile, for medical and food applications, Intervoid®
Lieferant: COVERIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
COV-112S COV-113S COV-114S COV-115S COV-116S COV-117S COV-118S COV-119S COV-120S COV-121S
129-1163EA 205 EUR InStock
129-1163 129-1164 129-1165 129-1166 129-1167 129-1168 129-1169 129-1170 129-1171 129-1172
Sampling bag, sterile, for medical and food applications, Intervoid®
Beutel Probenbeutel
The Intervoid® secure sampling bags are designed to protect and transport mainly critical samples ; they combine the highest levels of tamper evident security and tracking technologies to bring real benefits to multiple analytical processes.
  • High premium sterilization standards (10-6 gamma): bags closed on production, can be open by pre-cut under the security tape ensuring only the bag being used is exposed, greatly reducing wastage
  • Can be used both for primary or secondary packaging
  • Leak resistant
  • Single trip by design to avoid any risk of cross
More Product Information
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..