Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H131931000 F131930102 F131930500
129-4948EA 78.5 EUR InStock
129-4948 129-1475 129-1654
Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Gestelle Beutelständer
Economical and convenient, this bench top disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc during daily use.
  • Snug fitting polypropylene plastic cover helps hold the bag securely in place
  • Reusable, can be cleaned by hand or in automated dish washers
  • Autoclavable
The rectangular steel wire frame holds a 220×280 mm polyethylene bag which is coated in easy to clean, durable, orange colour epoxy.
Zertifizierungen: Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes.
Lieferumfang: Kit includes 100 bench top biohazard bags (129-4922); can also be used with Clavies autoclavable bags.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge