Moisture barrier bags, Dri-Shield® 2000 series
Lieferant: DESCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7001012 7001618
129-2645EA 61.7 EUR InStock
129-2645 129-2646
Moisture barrier bags, Dri-Shield® 2000 series
Beutel Aufbewahrungsbeutel
This aluminised moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area. The bags are heat sealable and suitable for vacuum packaging.
  • 0,0036 " (3.6 mil) thick bag film laminate constructed from multiple layers of aluminised polyester and polyethylene
  • Designed for moisture and electrostatic discharge (ESD) sensitive items (such as SMT)
  • <0,03 Grams Moisture Vapor Transmission Rate (MVTR); designed for dry packing of moisture sensitive devices and to protect contents from damage due to ESD events or EMI
  • <10 nJ Energy Penetration Tested per ANSI/ESD STM11.31; protects contents from ESD charges and discharges
  • >20lb puncture strength average; provides physical protection for contents
  • Dissipative Inner and Outer Surfaces Rs 1×10⁴ to < 1×10¹¹ Ω per ANSI/ESD STM11.11; allows bag to be grounded when placed on a grounded surface or handled by a grounded operator
Polyester provides puncture resistance. Metal layers are intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field while providing moisture protection.
Printed with ESD Protective Symbol and lot code information.
Zertifizierungen: Meets ANSI/ESD S20.20, Packaging standard ANSI/ESDS541, and Static Control Bag ANSI/ESD S11.4 Level 3 (except Transparency).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge