Lieferant: Flexico
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ITM031403 ITM031404 ITM031405 ITM031406 ITM031407 ITM031408 ITM031409 ITM031410 ITM031411 ITM031412 ITM031413 ITM031415 ITM031416 ITM031417 ITM031420 ITM031421 ITM031422 ITM031431 ITM031804
129-3910EA 182 EUR InStock
129-3910 129-3917 129-3918 129-3919 129-3920 129-3921 129-3922 129-3923 129-3924 129-3925 129-3926 129-3927 129-3928 129-3929 129-3930 129-3931 129-3932 129-3933 129-3934
Beutel Probenbeutel
The adhesive flap bag allows you to package your products manually and quickly. The bag is available with resealable or permanent adhesive. Flexico®’s adhesive flap bags are made from polypropylene or bioriented polypropylene in order to
ensure perfect transparency and highlight the products within.
The resealable adhesive flap bag makes it possible to test and touch the product without damaging the packaging.
*Bottom Gusset (BG)
Achtung! Bitte beachten: Keep out of reach of children
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge