Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Lieferant: VWR Seradigm
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SERD76419-586EA 71.2 EUR InStock
SERD76419-586 SERD76419-590 1300-500CR 1300-500CG 1300-500CGH 1300-500CH
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Seren
Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
  • Industry-leading collection techniques at the source
  • Risk mitigation through a robust quality system
  • Proven track record for performance and consistency
  • Sourced from countries with health status classified as “Negligible BSE Risk” by the World Health Organization
The care and attention we devote to the collection and manufacture of our products generates quality that matters. Avantor Seradigm Select Grade FBS preserves more of the naturally-occurring growth factors that cells require to thrive, resulting in the highest possible quality sera for a price that fits your budget.
Example Certificates of Analysis show typical results. Select Grade FBS is sourced from countries in Central America.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge