Stabilization:
1-5% Hexan
13060.14
13060.22
13060.88
13060.14EA
224
EUR
InStock
13060.14
13060.22
13060.88
Dicobalt octacarbonyl Stabilised
Dicobalt octacarbonyl
CAS No.: 10210-68-1
Molecular Formula: C8Co2O8
Formula Weight: 341.95
Flash Point: -13°(-9°F)
Physical Form: Crystalline
Density: 1.73
Sensitivity: Air Sensitive
Appearance: Red-orange
Melting Point: ca 51° dec.
Solubility: Insoluble in water. Soluble in alcohol, ether and carbon disulfide
UN No.: UN3124
MDL No.: MFCD00016024
Applications: As a catalyst in oxo process, and in high-purity cobalt salts