13076.18
13076.30
13076.18EA
46.1
EUR
InStock
13076.18
13076.30
Kupfer(II)fluorid Dihydrat
Kupfer(II)fluorid Dihydrat
CAS No.: 13454-88-1
Molecular Formula: CuF2.2H2O
Formula Weight: 137.57 (101.54anhy)
Physical Form: Powder
Density: 2.934
Odour: Odourless
Merck Reference: 14,2638
UN No.: 3288
MDL No.: MFCD00010977
Flux in casting gray iron