13198.18
13198.30
13198.09
13198.18EA
126
EUR
InStock
13198.18
13198.30
13198.09
Lanthanum(III) bromide hydrate 99,99
Lanthanum(III) bromide hydrate
Purity: 99.99%
CAS No.: 224183-16-8
Molecular Formula: LaBr3.xH2O
Formula Weight: 378.62(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00798151