Cleanroom tables, ergo
Lieferant: Palbam
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WX1201 WX1203 WX1401 WX1403 WX0604
132-0302EA 2390 EUR InStock
132-0302 132-0314 132-0304 132-0316 PALBWX0604
Cleanroom tables, ergo
Möbel Tische und Werkbänke
These cleanroom ergo tables are manufactured with 304 stainless steel with electropolished finish and Al anodized cleanroom compatible connectors.
  • Rounded front table tops
  • Excellent design conforms to highest cleanroom specifications
  • Ergonomic table legs designed to prevent operators 'knocking knees' on table
  • Height adjustable feet or casters
  • All accessories attachable
The bases are fabricated from tubing for easy cleaning and optimal airflow. The tops are designed to reduce vibrations and are very strong holding up to 80 kg.
Lieferumfang: The tables are shipped flat-packed with easy assembly instructions (assembly time ~10 minutes). Other shelves are available upon request.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge