13405.30
13405.14
13405.A1
13405.A7
13405.30EA
81.4
EUR
InStock
13405.30
13405.14
13405.A1
13405.A7
Lithiumnitrat, anhydrous 99%
Lithium nitrate
Purity: 99%
CAS No.: 7790-69-4
Molecular Formula: LiNO3
Formula Weight: 68.95
Physical Form: Crystalline
Density: 2.38
Appearance: White
Odour: Odourless
Melting Point: 264°
Boiling Point: 600° dec.
Solubility: Soluble in water, alcohol, NH4OH, and pyridine
Merck Reference: 14,5536
UN No.: 2722
MDL No.: MFCD00011094
Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant