13454.36
13454.A3
13454.A9
13454.36EA
41.7
EUR
InStock
13454.36
13454.A3
13454.A9
Natriumsulfit ≥98,0% ACS
Sodium sulphite
Purity: 98.0% min
CAS No.: 7757-83-7
Molecular Formula: Na2SO3
Formula Weight: 126.04
Physical Form: Granular
Density: 2.633
Appearance: White
Melting Point: 500° dec.
Solubility: Soluble in water, glycerol. Practically insoluble in alcohol
Merck Reference: 14,8682
MDL No.: MFCD00003503
Photographic developers, generating SO2, reducing agent (dyes), textile bleaching (antichlor)