13492.14
13492.22
13492.14EA
59.9
EUR
InStock
13492.14
13492.22
Cäsiumchromat 99,9 Metals basis
Cäsiumchromat
Purity: 99.9%
CAS No.: 13454-78-9
Molecular Formula: Cs2CrO4
Formula Weight: 381.81
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3087
MDL No.: MFCD00016050