Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Safety cabinets for dangerous products, Series 14

Lieferant: TRIONYX

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

ASDP
135-1680EA 951 EUR InStock
135-1680
Safety cabinets for dangerous products, Series 14
Möbel Lagerschränke
Safety cabinets for dangerous products, for the safe storage of dangerous products, in compliance with legislation.
Supplied with three standardised pictograms: 'Corrosive', 'Toxic' and 'Flammable' (to be affixed depending on use).

  • Shelves/containment trays with a height of 50 mm
  • Maximum 80 kg load per level (uniformly distributed)
  • Height-adjustable shelves/containment trays on rack with increment height of 15 mm

Made from a single piece of steel, epoxy paint: grey body (RAL 7035), yellow door(s) (RAL 1023).

Zertifizierungen: Complies with articles 5162 and 5170 of the French Public Health Code.
Complies with I.N.R.S. (Institut national de recherche et de sécurité - National Research and Safety Institute) recommendations.
Standardised pictograms comply with NF X08.003 and ISO 3864 standards, and with Council Directive 92/58/CEE.
G.S. certified.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge