Transport trolleys for lab
Lieferant: Metro International
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CHEMX3 LABX3 MDSX3
139-0059EA 1970 EUR InStock
139-0059 139-0075 139-0076
Transport trolleys for lab
Transportwagen
Metro’s Lab Cart line includes a General Lab Cart, Chemical Cart, and Mobile Desk. All carts are designed to offer superior corrosion resistance. Smooth rolling, 127 mm polyurethane castors come as standard with each cart. All shelves adjust in 12,7 mm increments, offering precise shelf adjustment and maximum storage utilisation. Microban antimicrobial protects the lower shelf mat from bacteria, mold, mildew and fungi that cause odors, stains and product degradation.
Mobile Desk:
Mobile Desk can be used for instrumentation storage or can be combined with accessories to provide a functional lab work surface. The rear posts are 1524 mm hieght to accommodate the monitor/utility bracket.
Chemical Cart:
Chemical Cart is designed to offer superior corrosion resistance in a lab environment. Stackable perimeter ledges retain chemicals, acid and other solution containers for safe transport. Solid shelf mats lift off of the shelf frame for easy cleaning.
General Lab Cart:
General Lab Cart is designed to offer superior corrosion resistance in a lab environment. Ideal for transporting equipment and instruments in and around the laboratory.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge